Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PUS7
Entrez Gene ID
54517
Gene Name
pseudouridylate synthase 7 (putative)
Gene Aliases
-
Chromosome Location
Chr.7: 105456501 - 105522270 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318163.1 NP_001305092.1 10 - 11 1262 122
NM_001318164.1 NP_001305093.1 10 - 11 1451 122
NM_019042.4 NP_061915.2 10 - 11 1475 122
XM_005250462.3 XP_005250519.1 11 - 12 1501 122
XM_011516336.2 XP_011514638.1 11 - 12 1496 122
XM_017012367.1 XP_016867856.1 11 - 12 1429 122
XM_017012368.1 XP_016867857.1 9 - 10 1342 122
GenBank mRNA AB067484.1 - 10 - 11 1189 122
AK091283.1 - 10 - 11 1455 122
AK128629.1 - 10 - 11 1437 122
BC005209.2 - 2 - 3 188 122
BC011396.1 - 10 - 11 1431 122

View Full Product Details