Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PUS7
Entrez Gene ID
54517
Gene Name
pseudouridylate synthase 7 (putative)
Gene Aliases
-
Chromosome Location
Chr.7: 105456501 - 105522270 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019042.4 NP_061915.2 1 - 2 209 118
XM_005250462.3 XP_005250519.1 1 - 2 217 118
XM_011516336.2 XP_011514638.1 1 - 2 212 118
XM_017012368.1 XP_016867857.1 1 - 2 221 118
GenBank mRNA AK091283.1 - 1 - 2 189 118
BC011396.1 - 1 - 2 165 118

View Full Product Details