Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PUS7
Entrez Gene ID
54517
Gene Name
pseudouridylate synthase 7 (putative)
Gene Aliases
-
Chromosome Location
Chr.7: 105456501 - 105522270 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318164.1 NP_001305093.1 5 - 6 948 75
NM_019042.4 NP_061915.2 5 - 6 972 75
GenBank mRNA AK091283.1 - 5 - 6 952 75
BC011396.1 - 5 - 6 928 75

View Full Product Details