Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1orf35
Entrez Gene ID
79169
Gene Name
chromosome 1 open reading frame 35
Gene Aliases
MMTAG2
Chromosome Location
Chr.1: 228100727 - 228103462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024319.3 NP_077295.1 7 - 8 905 62
NR_130975.1 8 - 9 1085 62
GenBank mRNA AK123377.1 - 4 - 5 1730 62
AK126087.1 - 8 - 9 3034 62
AY137773.1 - 7 - 8 766 62
BC002843.2 - 7 - 8 764 62
BC062585.1 - 6 - 7 905 62

View Full Product Details