Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCA
Entrez Gene ID
25801
Gene Name
grancalcin
Gene Aliases
GCL
Chromosome Location
Chr.2: 162318768 - 162371605 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012198.3 NP_036330.1 5 - 6 629 135
XM_005246446.2 XP_005246503.1 5 - 6 570 135
XM_006712398.3 XP_006712461.1 5 - 6 802 135
XM_006712400.3 XP_006712463.1 6 - 7 880 135
XM_006712401.3 XP_006712464.1 5 - 6 881 135
XM_011510926.2 XP_011509228.1 5 - 6 581 135
XM_011510927.2 XP_011509229.1 5 - 6 576 135
XM_011510928.2 XP_011509230.1 3 - 4 514 135
XM_017003764.1 XP_016859253.1 6 - 7 893 135
XM_017003765.1 XP_016859254.1 6 - 7 979 135
XM_017003766.1 XP_016859255.1 6 - 7 776 135
XM_017003767.1 XP_016859256.1 5 - 6 652 135
XM_017003768.1 XP_016859257.1 5 - 6 647 135
GenBank mRNA AB209768.1 - 5 - 6 518 135
AK295413.1 - 5 - 6 562 135
AK312349.1 - 5 - 6 578 135
BC005214.1 - 5 - 6 552 135
M81637.1 - 5 - 6 572 135

View Full Product Details