The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042355.1 | NP_001035814.1 | 2 - 3 | 157 | 75 |
NM_001042412.1 | NP_001035877.1 | 2 - 3 | 181 | 75 | |
XM_011522908.1 | XP_011521210.1 | 2 - 3 | 172 | 75 | |
XM_011522911.2 | XP_011521213.1 | 2 - 3 | 172 | 75 | |
XM_017022977.1 | XP_016878466.1 | 2 - 3 | 452 | 75 | |
XM_017022981.1 | XP_016878470.1 | 2 - 3 | 139 | 75 | |
GenBank mRNA | AK056226.1 | - | 2 - 3 | 112 | 75 |
AK292975.1 | - | 2 - 3 | 139 | 75 | |
BC012342.1 | - | 2 - 3 | 155 | 75 | |
DA785459.1 | - | 2 - 3 | 157 | 75 |