The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042355.1 | NP_001035814.1 | 6 - 7 | 1290 | 119 |
NM_001042412.1 | NP_001035877.1 | 6 - 7 | 1314 | 119 | |
NM_015247.2 | NP_056062.1 | 8 - 9 | 1436 | 119 | |
XM_005255812.2 | XP_005255869.1 | 7 - 8 | 1387 | 119 | |
XM_006721149.1 | XP_006721212.1 | 7 - 8 | 1451 | 119 | |
XM_011522907.2 | XP_011521209.1 | 6 - 7 | 1213 | 119 | |
XM_011522908.1 | XP_011521210.1 | 6 - 7 | 1305 | 119 | |
XM_011522911.2 | XP_011521213.1 | 6 - 7 | 1305 | 119 | |
XM_017022977.1 | XP_016878466.1 | 6 - 7 | 1585 | 119 | |
XM_017022978.1 | XP_016878467.1 | 6 - 7 | 1381 | 119 | |
XM_017022979.1 | XP_016878468.1 | 6 - 7 | 1341 | 119 | |
XM_017022980.1 | XP_016878469.1 | 6 - 7 | 1374 | 119 | |
XM_017022981.1 | XP_016878470.1 | 6 - 7 | 1272 | 119 | |
GenBank mRNA | AJ250014.1 | - | 8 - 9 | 1412 | 119 |
AK000187.1 | - | 1 - 2 | 92 | 119 | |
AK024212.1 | - | 4 - 5 | 748 | 119 | |
AK024348.1 | - | 4 - 5 | 797 | 119 | |
AK056226.1 | - | 6 - 7 | 1245 | 119 | |
AK225202.1 | - | 7 - 8 | 1407 | 119 | |
AK292975.1 | - | 6 - 7 | 1272 | 119 | |
AK310832.1 | - | 6 - 7 | 1262 | 119 | |
BC012342.1 | - | 6 - 7 | 1288 | 119 |