Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRCAM
Entrez Gene ID
4897
Gene Name
neuronal cell adhesion molecule
Gene Aliases
-
Chromosome Location
Chr.7: 108147626 - 108456442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037132.2 NP_001032209.1 18 - 19 2336 110
NM_001193582.1 NP_001180511.1 21 - 22 2716 110
NM_001193583.1 NP_001180512.1 20 - 21 2659 110
NM_001193584.1 NP_001180513.1 20 - 21 2659 110
NM_005010.4 NP_005001.3 19 - 20 2668 110
XM_005250373.2 XP_005250430.1 21 - 22 2716 110
XM_005250383.2 XP_005250440.1 21 - 22 2716 110
XM_005250385.2 XP_005250442.1 21 - 22 2716 110
XM_006716003.1 XP_006716066.1 21 - 22 2716 110
XM_006716007.1 XP_006716070.1 21 - 22 2716 110
XM_006716012.1 XP_006716075.1 21 - 22 2716 110
XM_006716014.1 XP_006716077.1 21 - 22 2716 110
XM_011516253.1 XP_011514555.1 21 - 22 2716 110
XM_011516255.1 XP_011514557.1 20 - 21 2471 110
XM_011516256.1 XP_011514558.1 21 - 22 2716 110
XM_011516257.1 XP_011514559.1 20 - 21 2698 110
XM_011516258.1 XP_011514560.1 19 - 20 2453 110
XM_011516259.1 XP_011514561.1 20 - 21 2686 110
XM_011516261.1 XP_011514563.1 19 - 20 2668 110
XM_011516262.1 XP_011514564.1 20 - 21 2659 110
XM_011516265.2 XP_011514567.1 21 - 22 2716 110
XM_011516266.2 XP_011514568.1 21 - 22 2716 110
XM_011516267.1 XP_011514569.1 21 - 22 2716 110
XM_011516268.2 XP_011514570.1 19 - 20 2531 110
XM_011516269.1 XP_011514571.1 21 - 22 2716 110
XM_011516270.1 XP_011514572.1 21 - 22 2716 110
XM_011516271.1 XP_011514573.1 19 - 20 2668 110
XM_017012236.1 XP_016867725.1 20 - 21 2686 110
XM_017012237.1 XP_016867726.1 21 - 22 2716 110
XM_017012238.1 XP_016867727.1 20 - 21 2659 110
XM_017012239.1 XP_016867728.1 19 - 20 2641 110
XM_017012240.1 XP_016867729.1 20 - 21 2659 110
XM_017012241.1 XP_016867730.1 19 - 20 2383 110
XM_017012242.1 XP_016867731.1 19 - 20 2383 110
XM_017012243.1 XP_016867732.1 21 - 22 2716 110
XM_017012244.1 XP_016867733.1 21 - 22 2716 110
XM_017012245.1 XP_016867734.1 21 - 22 2716 110
XM_017012246.1 XP_016867735.1 20 - 21 2686 110
XM_017012247.1 XP_016867736.1 20 - 21 2659 110
XM_017012248.1 XP_016867737.1 20 - 21 2686 110
XM_017012249.1 XP_016867738.1 20 - 21 2659 110
XM_017012250.1 XP_016867739.1 19 - 20 2383 110
XM_017012251.1 XP_016867740.1 20 - 21 2686 110
XM_017012252.1 XP_016867741.1 20 - 21 2686 110
XM_017012253.1 XP_016867742.1 18 - 19 2385 110
XM_017012254.1 XP_016867743.1 20 - 21 2686 110
XM_017012255.1 XP_016867744.1 18 - 19 2426 110
XM_017012256.1 XP_016867745.1 18 - 19 2385 110
XM_017012257.1 XP_016867746.1 19 - 20 2414 110
XM_017012258.1 XP_016867747.1 19 - 20 2410 110
XM_017012259.1 XP_016867748.1 17 - 18 2355 110
GenBank mRNA AB002341.1 - 20 - 21 2587 110
AJ001057.1 - 18 - 19 2215 110
AK299870.1 - 21 - 22 2704 110
BC098401.1 - 19 - 20 2656 110
BC114570.1 - 20 - 21 2583 110
BC115736.1 - 20 - 21 2583 110
BX538010.1 - 19 - 20 2613 110

View Full Product Details