Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRCAM
Entrez Gene ID
4897
Gene Name
neuronal cell adhesion molecule
Gene Aliases
-
Chromosome Location
Chr.7: 108147626 - 108456442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037132.2 NP_001032209.1 6 - 7 825 101
NM_001193582.1 NP_001180511.1 9 - 10 1205 101
NM_005010.4 NP_005001.3 8 - 9 1187 101
XM_005250373.2 XP_005250430.1 9 - 10 1205 101
XM_005250383.2 XP_005250440.1 9 - 10 1205 101
XM_005250385.2 XP_005250442.1 9 - 10 1205 101
XM_006716003.1 XP_006716066.1 9 - 10 1205 101
XM_006716007.1 XP_006716070.1 9 - 10 1205 101
XM_006716012.1 XP_006716075.1 9 - 10 1205 101
XM_006716014.1 XP_006716077.1 9 - 10 1205 101
XM_011516253.1 XP_011514555.1 9 - 10 1205 101
XM_011516255.1 XP_011514557.1 8 - 9 960 101
XM_011516256.1 XP_011514558.1 9 - 10 1205 101
XM_011516257.1 XP_011514559.1 8 - 9 1187 101
XM_011516258.1 XP_011514560.1 7 - 8 942 101
XM_011516259.1 XP_011514561.1 9 - 10 1205 101
XM_011516261.1 XP_011514563.1 8 - 9 1187 101
XM_011516265.2 XP_011514567.1 9 - 10 1205 101
XM_011516266.2 XP_011514568.1 9 - 10 1205 101
XM_011516267.1 XP_011514569.1 9 - 10 1205 101
XM_011516268.2 XP_011514570.1 7 - 8 1020 101
XM_011516269.1 XP_011514571.1 9 - 10 1205 101
XM_011516270.1 XP_011514572.1 9 - 10 1205 101
XM_011516271.1 XP_011514573.1 8 - 9 1187 101
XM_017012236.1 XP_016867725.1 9 - 10 1205 101
XM_017012237.1 XP_016867726.1 9 - 10 1205 101
XM_017012243.1 XP_016867732.1 9 - 10 1205 101
XM_017012244.1 XP_016867733.1 9 - 10 1205 101
XM_017012245.1 XP_016867734.1 9 - 10 1205 101
XM_017012246.1 XP_016867735.1 9 - 10 1205 101
XM_017012248.1 XP_016867737.1 9 - 10 1205 101
XM_017012251.1 XP_016867740.1 9 - 10 1205 101
XM_017012252.1 XP_016867741.1 9 - 10 1205 101
XM_017012253.1 XP_016867742.1 6 - 7 874 101
XM_017012254.1 XP_016867743.1 9 - 10 1205 101
XM_017012256.1 XP_016867745.1 6 - 7 874 101
XM_017012258.1 XP_016867747.1 8 - 9 929 101
XM_017012259.1 XP_016867748.1 6 - 7 874 101
GenBank mRNA AJ001057.1 - 6 - 7 704 101
AK299870.1 - 9 - 10 1193 101
BC098401.1 - 8 - 9 1175 101
BX538010.1 - 8 - 9 1132 101

View Full Product Details