Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC9A3R1
Entrez Gene ID
9368
Gene Name
SLC9A3 regulator 1
Gene Aliases
EBP50, NHERF, NHERF-1, NHERF1, NPHLOP2
Chromosome Location
Chr.17: 74748612 - 74769360 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004252.4 NP_004243.1 4 - 5 1026 62
GenBank mRNA AF015926.1 - 4 - 5 1003 62
AF036241.1 - 4 - 5 961 62
AK026581.1 - 4 - 5 1009 62
AK128474.1 - 5 - 6 975 62
AK312498.1 - 4 - 5 1042 62
BC001443.2 - 4 - 5 979 62
BC003361.1 - 4 - 5 996 62
BC011777.1 - 4 - 5 995 62
BC049220.1 - 4 - 5 1002 62
BC053350.1 - 4 - 5 986 62
BX648303.1 - 2 - 3 2685 62

View Full Product Details