The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258012.3 | NP_001244941.2 | 6 - 7 | 825 | 62 |
NM_001258013.3 | NP_001244942.2 | 7 - 8 | 789 | 62 | |
NM_001258014.3 | NP_001244943.2 | 6 - 7 | 672 | 62 | |
NM_001258015.2 | NP_001244944.1 | 5 - 6 | 576 | 62 | |
NM_001258016.3 | NP_001244945.2 | 5 - 6 | 641 | 62 | |
NM_024417.4 | NP_077728.3 | 6 - 7 | 696 | 62 | |
NR_047576.2 | 6 - 7 | 796 | 62 | ||
XM_006721772.1 | XP_006721835.1 | 6 - 7 | 644 | 62 | |
XM_011524533.2 | XP_011522835.1 | 5 - 6 | 622 | 62 | |
GenBank mRNA | AK094120.1 | - | 7 - 8 | 894 | 62 |
AK098648.1 | - | 6 - 7 | 633 | 62 | |
AK223530.1 | - | 6 - 7 | 626 | 62 | |
AK293208.1 | - | 7 - 8 | 734 | 62 | |
AK293210.1 | - | 6 - 7 | 733 | 62 | |
AK295307.1 | - | 6 - 7 | 825 | 62 | |
AK298908.1 | - | 5 - 6 | 519 | 62 | |
AK301779.1 | - | 6 - 7 | 609 | 62 | |
AK301977.1 | - | 5 - 6 | 641 | 62 | |
BC002960.1 | - | 6 - 7 | 635 | 62 | |
BC018911.2 | - | 6 - 7 | 642 | 62 | |
BC034763.1 | - | 6 - 7 | 1297 | 62 | |
BC063493.1 | - | 6 - 7 | 634 | 62 | |
BT006684.1 | - | 6 - 7 | 609 | 62 |