The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133637.2 | NP_598376.2 | 6 - 7 | 1307 | 90 |
XM_011532644.1 | XP_011530946.1 | 5 - 6 | 986 | 90 | |
XM_011532645.1 | XP_011530947.1 | 3 - 4 | 595 | 90 | |
XM_017003520.1 | XP_016859009.1 | 3 - 4 | 851 | 90 | |
GenBank mRNA | AK074337.1 | - | 6 - 7 | 1214 | 90 |
BC075018.2 | - | 4 - 5 | 834 | 90 |