Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DQX1
Entrez Gene ID
165545
Gene Name
DEAQ-box RNA dependent ATPase 1
Gene Aliases
-
Chromosome Location
Chr.2: 74518131 - 74526281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133637.2 NP_598376.2 7 - 8 1467 105
XM_011532644.1 XP_011530946.1 6 - 7 1146 105
XM_011532645.1 XP_011530947.1 4 - 5 755 105
XM_017003520.1 XP_016859009.1 4 - 5 1011 105
GenBank mRNA AK074337.1 - 7 - 8 1374 105
BC075018.2 - 5 - 6 994 105

View Full Product Details