The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168474.1 | NP_001161946.1 | 1 - 2 | 112 | 93 |
XM_005262152.1 | XP_005262209.1 | 1 - 2 | 112 | 93 | |
GenBank mRNA | AF285595.1 | - | 1 - 2 | 114 | 93 |
DB042169.1 | - | 1 - 2 | 112 | 93 |