The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012397.3 | NP_036529.1 | 2 - 3 | 334 | 141 |
GenBank mRNA | AF169949.1 | - | 1 - 2 | 196 | 141 |
AJ001696.2 | - | 2 - 3 | 268 | 141 | |
AJ001697.2 | - | 2 - 3 | 236 | 141 | |
AK290323.1 | - | 2 - 3 | 308 | 141 | |
AK313486.1 | - | 2 - 3 | 306 | 141 | |
BC101821.1 | - | 2 - 3 | 223 | 141 |