The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173642.3 | NP_775913.2 | 2 - 3 | 533 | 70 |
XM_006710585.3 | XP_006710648.1 | 2 - 3 | 527 | 70 | |
XM_017001082.1 | XP_016856571.1 | 3 - 4 | 592 | 70 | |
GenBank mRNA | BC039737.1 | - | 2 - 3 | 527 | 70 |
DR000262.1 | - | 3 - 4 | 603 | 69 |