The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001309192.1 | NP_001296121.1 | 10 - 11 | 2321 | 91 |
NM_001309193.1 | NP_001296122.1 | 12 - 13 | 2495 | 91 | |
NM_004442.7 | NP_004433.2 | 12 - 13 | 2498 | 91 | |
NM_017449.4 | NP_059145.2 | 12 - 13 | 2495 | 91 | |
XM_006710441.3 | XP_006710504.1 | 12 - 13 | 2591 | 91 | |
XM_006710442.3 | XP_006710505.1 | 12 - 13 | 2473 | 91 | |
GenBank mRNA | AB210018.1 | - | 12 - 13 | 2404 | 91 |
AF025304.1 | - | 12 - 13 | 2375 | 91 | |
AK298898.1 | - | 8 - 9 | 1172 | 91 | |
AK299701.1 | - | 11 - 12 | 2260 | 91 | |
BC007903.2 | - | 11 - 12 | 2273 | 91 | |
BC018763.1 | - | 10 - 11 | 1648 | 91 | |
D14717.1 | - | 2 - 3 | 433 | 91 | |
D31661.1 | - | 12 - 13 | 2359 | 91 | |
D37827.1 | - | 6 - 7 | 826 | 91 | |
L36643.1 | - | 12 - 13 | 91 | ||
L41939.1 | - | 12 - 13 | 2457 | 91 |