The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001309192.1 | NP_001296121.1 | 2 - 3 | 273 | 73 |
NM_001309193.1 | NP_001296122.1 | 2 - 3 | 273 | 73 | |
NM_004442.7 | NP_004433.2 | 2 - 3 | 273 | 73 | |
NM_017449.4 | NP_059145.2 | 2 - 3 | 273 | 73 | |
XM_006710441.3 | XP_006710504.1 | 2 - 3 | 366 | 73 | |
XM_006710442.3 | XP_006710505.1 | 2 - 3 | 341 | 73 | |
GenBank mRNA | AB210018.1 | - | 2 - 3 | 182 | 73 |
AF025304.1 | - | 2 - 3 | 153 | 73 | |
BC007903.2 | - | 1 - 2 | 51 | 73 | |
BC067861.1 | - | 2 - 3 | 272 | 73 | |
D31661.1 | - | 2 - 3 | 134 | 73 | |
DB282024.1 | - | 3 - 4 | 214 | 73 | |
L36643.1 | - | 2 - 3 | 86 | 73 | |
L41939.1 | - | 2 - 3 | 232 | 73 |