Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VWA5B1
Entrez Gene ID
127731
Gene Name
von Willebrand factor A domain containing 5B1
Gene Aliases
-
Chromosome Location
Chr.1: 20290907 - 20360442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039500.2 NP_001034589.2 15 - 16 2510 70
XM_006710360.3 XP_006710423.2 12 - 13 1785 70
XM_011540683.2 XP_011538985.1 15 - 16 2472 70
XM_011540684.2 XP_011538986.1 15 - 16 2597 70
XM_011540685.2 XP_011538987.1 15 - 16 2609 70
XM_011540688.2 XP_011538990.1 15 - 16 2472 70
XM_011540690.2 XP_011538992.1 14 - 15 2295 70
XM_011540691.2 XP_011538993.1 14 - 15 2286 70
XM_011540692.2 XP_011538994.1 15 - 16 2466 70
XM_011540693.1 XP_011538995.1 15 - 16 2472 70
XM_011540695.1 XP_011538997.1 15 - 16 2472 70
XM_011540697.2 XP_011538999.1 9 - 10 1843 70
XM_017000305.1 XP_016855794.1 9 - 10 1589 70
XM_017000306.1 XP_016855795.1 10 - 11 1750 70
GenBank mRNA AK057346.1 - 4 - 5 593 70
AK295797.1 - 15 - 16 2494 70
BC101378.1 - 4 - 5 590 70
BC101379.2 - 4 - 5 590 70
BC101380.1 - 4 - 5 590 70
BC101381.1 - 4 - 5 590 70

View Full Product Details