The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197259.1 | NP_001184188.1 | 11 - 12 | 1299 | 110 |
NM_012381.3 | NP_036513.2 | 12 - 13 | 1397 | 110 | |
NM_181837.2 | NP_862820.1 | 12 - 13 | 1397 | 110 | |
XM_005248704.1 | XP_005248761.1 | 12 - 13 | 1343 | 110 | |
XM_005248705.1 | XP_005248762.1 | 13 - 14 | 1401 | 110 | |
XM_005248706.1 | XP_005248763.1 | 13 - 14 | 1400 | 110 | |
XM_011535651.1 | XP_011533953.1 | 11 - 12 | 1229 | 110 | |
XM_011535652.2 | XP_011533954.1 | 12 - 13 | 1343 | 110 | |
XM_017010632.1 | XP_016866121.1 | 12 - 13 | 1343 | 110 | |
XM_017010633.1 | XP_016866122.1 | 12 - 13 | 1343 | 110 | |
XM_017010634.1 | XP_016866123.1 | 12 - 13 | 1343 | 110 | |
XM_017010635.1 | XP_016866124.1 | 11 - 12 | 1229 | 110 | |
XM_017010636.1 | XP_016866125.1 | 13 - 14 | 1401 | 110 | |
XM_017010637.1 | XP_016866126.1 | 13 - 14 | 1400 | 110 | |
XM_017010638.1 | XP_016866127.1 | 13 - 14 | 1401 | 110 | |
XM_017010639.1 | XP_016866128.1 | 13 - 14 | 1400 | 110 | |
XM_017010640.1 | XP_016866129.1 | 12 - 13 | 1286 | 110 | |
GenBank mRNA | AF093535.1 | - | 12 - 13 | 1321 | 110 |
AF125507.1 | - | 12 - 13 | 1330 | 110 | |
AF135044.1 | - | 12 - 13 | 1295 | 110 | |
AK222795.1 | - | 12 - 13 | 1323 | 110 | |
AK302750.1 | - | 13 - 14 | 1391 | 110 | |
AK303056.1 | - | 12 - 13 | 1323 | 110 | |
AK303182.1 | - | 11 - 12 | 1118 | 110 | |
AK303195.1 | - | 11 - 12 | 1223 | 110 | |
AK315911.1 | - | 12 - 13 | 1284 | 110 | |
AK316298.1 | - | 12 - 13 | 1328 | 110 | |
AL080116.1 | - | 12 - 13 | 1336 | 110 | |
BC035494.2 | - | 12 - 13 | 1312 | 110 | |
BC047689.1 | - | 12 - 13 | 1313 | 110 | |
CR456814.1 | - | 7 - 8 | 830 | 110 | |
U50950.1 | - | 8 - 9 | 963 | 110 |