The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197259.1 | NP_001184188.1 | 14 - 15 | 1599 | 157 |
NM_012381.3 | NP_036513.2 | 15 - 16 | 1697 | 157 | |
NM_181837.2 | NP_862820.1 | 15 - 16 | 1700 | 157 | |
XM_005248704.1 | XP_005248761.1 | 15 - 16 | 1646 | 157 | |
XM_005248705.1 | XP_005248762.1 | 16 - 17 | 1704 | 157 | |
XM_005248706.1 | XP_005248763.1 | 16 - 17 | 1703 | 157 | |
XM_011535651.1 | XP_011533953.1 | 14 - 15 | 1532 | 157 | |
XM_017010632.1 | XP_016866121.1 | 15 - 16 | 1646 | 157 | |
XM_017010633.1 | XP_016866122.1 | 15 - 16 | 1643 | 157 | |
XM_017010634.1 | XP_016866123.1 | 15 - 16 | 1643 | 157 | |
XM_017010635.1 | XP_016866124.1 | 14 - 15 | 1529 | 157 | |
XM_017010636.1 | XP_016866125.1 | 16 - 17 | 1701 | 157 | |
XM_017010637.1 | XP_016866126.1 | 16 - 17 | 1700 | 157 | |
XM_017010638.1 | XP_016866127.1 | 16 - 17 | 1701 | 157 | |
XM_017010639.1 | XP_016866128.1 | 16 - 17 | 1700 | 157 | |
XM_017010640.1 | XP_016866129.1 | 15 - 16 | 1586 | 157 | |
GenBank mRNA | AF093535.1 | - | 15 - 16 | 1621 | 157 |
AF125507.1 | - | 15 - 16 | 1630 | 157 | |
AF135044.1 | - | 15 - 16 | 1595 | 157 | |
AK222795.1 | - | 15 - 16 | 1623 | 157 | |
AK302750.1 | - | 17 - 18 | 1914 | 157 | |
AK303182.1 | - | 14 - 15 | 1418 | 157 | |
AK303195.1 | - | 14 - 15 | 1523 | 157 | |
AK315911.1 | - | 15 - 16 | 1584 | 157 | |
BC035494.2 | - | 15 - 16 | 1612 | 157 | |
BC047689.1 | - | 15 - 16 | 1616 | 157 | |
U50950.1 | - | 11 - 12 | 1263 | 157 |