The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256394.1 | NP_001243323.1 | 5 - 6 | 723 | 253 |
NM_017948.5 | NP_060418.4 | 4 - 5 | 619 | 137 | |
NR_046106.1 | 4 - 5 | 619 | 137 | ||
XM_006717166.3 | XP_006717229.1 | 4 - 5 | 485 | 137 | |
XM_006717167.3 | XP_006717230.1 | 4 - 5 | 607 | 137 | |
XM_006717168.3 | XP_006717231.1 | 4 - 5 | 378 | 137 | |
XM_006717169.3 | XP_006717232.1 | 5 - 6 | 638 | 253 | |
XM_006717170.3 | XP_006717233.1 | 5 - 6 | 506 | 253 | |
XM_011518824.2 | XP_011517126.1 | 3 - 4 | 203 | 137 | |
XM_017014876.1 | XP_016870365.1 | 4 - 5 | 485 | 137 | |
XM_017014877.1 | XP_016870366.1 | 5 - 6 | 506 | 253 | |
GenBank mRNA | AB105104.1 | - | 4 - 5 | 585 | 137 |
AB109030.1 | - | 3 - 4 | 282 | 137 | |
AK001049.1 | - | 4 - 5 | 501 | 137 | |
AK297355.1 | - | 4 - 5 | 502 | 137 | |
AK307408.1 | - | 4 - 5 | 510 | 137 | |
BC146810.1 | - | 5 - 6 | 479 | 253 | |
CR627363.1 | - | 4 - 5 | 363 | 137 |