Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCAF11
Entrez Gene ID
80344
Gene Name
DDB1 and CUL4 associated factor 11
Gene Aliases
GL014, PRO2389, WDR23
Chromosome Location
Chr.14: 24114697 - 24125242 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163484.1 NP_001156956.1 6 - 7 853 86
NM_025230.4 NP_079506.3 6 - 7 1303 86
NM_181357.2 NP_852002.1 6 - 7 655 86
NR_028099.1 5 - 6 1215 86
NR_028100.1 5 - 6 830 86
GenBank mRNA AB209073.1 - 5 - 6 1201 86
AF130070.1 - 5 - 6 563 86
AF267858.1 - 6 - 7 966 86
AK057636.1 - 6 - 7 614 86
AK074296.1 - 6 - 7 1268 86
AK094478.1 - 5 - 6 432 86
AK292797.1 - 6 - 7 735 86
AK296742.1 - 5 - 6 787 86
BC008858.2 - 1 - 2 81 86
BC067132.1 - 6 - 7 726 86
BX161405.1 - 5 - 6 550 86
BX247970.1 - 4 - 5 729 86
BX648224.1 - 6 - 7 785 86

View Full Product Details