Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNB3
Entrez Gene ID
85417
Gene Name
cyclin B3
Gene Aliases
-
Chromosome Location
Chr.X: 50202713 - 50351914 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033031.2 NP_149020.2 5 - 6 3444 68
XM_017029916.1 XP_016885405.1 6 - 7 3628 68
XM_017029918.1 XP_016885407.1 6 - 7 3628 68
GenBank mRNA AJ314764.1 - 5 - 6 3448 68
AJ416458.1 - 5 - 6 3460 68
AK292454.1 - 6 - 7 3617 68

View Full Product Details