The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323370.1 | NP_001310299.1 | 3 - 4 | 618 | 130 |
NM_003881.3 | NP_003872.1 | 2 - 3 | 822 | 130 | |
GenBank mRNA | AF074604.1 | - | 2 - 3 | 281 | 130 |
AF083500.1 | - | 2 - 3 | 283 | 130 | |
AF100780.1 | - | 2 - 3 | 430 | 130 | |
AK313853.1 | - | 2 - 3 | 298 | 130 | |
AY358915.1 | - | 2 - 3 | 284 | 130 | |
BC017782.2 | - | 2 - 3 | 421 | 130 | |
BC064379.1 | - | 2 - 3 | 285 | 130 |