The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015041.2 | NP_055856.1 | 3 - 4 | 344 | 111 |
XM_006720867.3 | XP_006720930.1 | 2 - 3 | 195 | 111 | |
XM_017023068.1 | XP_016878557.1 | 3 - 4 | 359 | 111 | |
XM_017023069.1 | XP_016878558.1 | 3 - 4 | 359 | 111 | |
XM_017023070.1 | XP_016878559.1 | 3 - 4 | 344 | 111 | |
XM_017023071.1 | XP_016878560.1 | 3 - 4 | 1333 | 111 | |
XM_017023072.1 | XP_016878561.1 | 3 - 4 | 1333 | 111 | |
XM_017023073.1 | XP_016878562.1 | 2 - 3 | 195 | 111 | |
GenBank mRNA | AB014543.1 | - | 3 - 4 | 271 | 111 |
AB089691.1 | - | 3 - 4 | 326 | 111 | |
BC017070.2 | - | 3 - 4 | 272 | 111 | |
DA502615.1 | - | 3 - 4 | 347 | 111 |