The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006052.1 | NP_006043.1 | 7 - 8 | 1045 | 70 |
XM_005260909.4 | XP_005260966.1 | 6 - 7 | 1555 | 70 | |
XM_005260911.4 | XP_005260968.1 | 4 - 5 | 1261 | 70 | |
XM_011529423.2 | XP_011527725.1 | 6 - 7 | 1492 | 70 | |
GenBank mRNA | AK299662.1 | - | 5 - 6 | 603 | 70 |
AK300024.1 | - | 6 - 7 | 821 | 70 | |
AK303624.1 | - | 4 - 5 | 692 | 70 | |
AK312707.1 | - | 7 - 8 | 1000 | 70 | |
AK316460.1 | - | 6 - 7 | 737 | 70 | |
BC110655.1 | - | 7 - 8 | 1487 | 70 | |
BX648191.1 | - | 9 - 10 | 2187 | 70 | |
D87343.1 | - | 7 - 8 | 1045 | 70 |