Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HK1
Entrez Gene ID
3098
Gene Name
hexokinase 1
Gene Aliases
HK, HK1-ta, HK1-tb, HK1-tc, HKD, HKI, HMSNR, HXK1, hexokinase
Chromosome Location
Chr.10: 69270000 - 69401882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000188.2 NP_000179.2 8 - 9 1128 63
NM_001322364.1 NP_001309293.1 10 - 11 1221 63
NM_001322365.1 NP_001309294.1 13 - 14 1526 63
NM_001322366.1 NP_001309295.1 8 - 9 1019 63
NM_001322367.1 NP_001309296.1 7 - 8 1032 63
NM_033496.2 NP_277031.1 8 - 9 1125 63
NM_033497.2 NP_277032.1 11 - 12 1343 63
NM_033498.2 NP_277033.1 11 - 12 1397 63
NM_033500.2 NP_277035.2 12 - 13 1490 63
XM_005269737.1 XP_005269794.1 8 - 9 1399 63
XM_011539732.1 XP_011538034.1 12 - 13 1397 63
GenBank mRNA AB209526.1 - 8 - 9 1124 63
AK091267.1 - 8 - 9 1253 63
AK128226.1 - 13 - 14 1521 63
AK292012.1 - 8 - 9 1128 63
AK294428.1 - 8 - 9 1125 63
AK309904.1 - 7 - 8 954 63
BC008730.2 - 8 - 9 1083 63
M75126.1 - 8 - 9 1108 63
X66957.1 - 2 - 3 169 63

View Full Product Details