The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001067.3 | NP_001058.2 | 11 - 12 | 1505 | 76 |
XM_005257632.1 | XP_005257689.1 | 11 - 12 | 1447 | 76 | |
XM_011525165.1 | XP_011523467.1 | 11 - 12 | 1451 | 76 | |
GenBank mRNA | AF285157.1 | - | 11 - 12 | 1424 | 76 |
AF285158.1 | - | 11 - 12 | 1454 | 76 | |
AF285159.1 | - | 11 - 12 | 1589 | 76 | |
BC013429.1 | - | 11 - 12 | 1472 | 76 | |
BC140791.1 | - | 11 - 12 | 1375 | 76 | |
J04088.1 | - | 11 - 12 | 1382 | 76 |