The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080878.2 | NP_543154.1 | 6 - 7 | 779 | 94 |
XM_011509210.1 | XP_011507512.1 | 8 - 9 | 1256 | 94 | |
XM_011509211.1 | XP_011507513.1 | 8 - 9 | 1253 | 94 | |
GenBank mRNA | AY065973.1 | - | 6 - 7 | 751 | 94 |
AY358905.1 | - | 6 - 7 | 779 | 94 | |
BC117225.1 | - | 6 - 7 | 777 | 94 | |
BC143339.1 | - | 5 - 6 | 791 | 94 | |
BC143341.1 | - | 6 - 7 | 777 | 94 |