Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH11
Entrez Gene ID
8701
Gene Name
dynein axonemal heavy chain 11
Gene Aliases
CILD7, DNAHBL, DNAHC11, DNHBL, DPL11
Chromosome Location
Chr.7: 21543215 - 21901568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277115.1 NP_001264044.1 64 - 65 10594 67
GenBank mRNA AJ320497.1 - 64 - 65 10615 67

View Full Product Details