Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTCD2
Entrez Gene ID
79810
Gene Name
pentatricopeptide repeat domain 2
Gene Aliases
-
Chromosome Location
Chr.5: 72320367 - 72368395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284403.1 NP_001271332.1 5 - 6 507 116
NM_001284404.1 NP_001271333.1 6 - 7 637 116
NM_001284405.1 NP_001271334.1 7 - 8 755 116
NM_024754.4 NP_079030.3 8 - 9 834 116
XM_005248603.2 XP_005248660.1 7 - 8 690 116
XM_017009866.1 XP_016865355.1 7 - 8 695 116
XM_017009867.1 XP_016865356.1 6 - 7 607 116
GenBank mRNA AK022660.1 - 6 - 7 596 116
AK056761.1 - 8 - 9 1498 116
AK298750.1 - 5 - 6 498 116
AK303639.1 - 6 - 7 606 116
AK303776.1 - 7 - 8 754 116
BC018720.1 - 8 - 9 810 116
CR457290.1 - 4 - 5 362 116

View Full Product Details