Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTCD2
Entrez Gene ID
79810
Gene Name
pentatricopeptide repeat domain 2
Gene Aliases
-
Chromosome Location
Chr.5: 72320367 - 72368395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284403.1 NP_001271332.1 6 - 7 635 139
NM_001284404.1 NP_001271333.1 7 - 8 765 139
NM_001284405.1 NP_001271334.1 8 - 9 883 139
NM_024754.4 NP_079030.3 9 - 10 962 139
XM_005248603.2 XP_005248660.1 8 - 9 818 139
XM_017009866.1 XP_016865355.1 8 - 9 823 139
XM_017009867.1 XP_016865356.1 7 - 8 735 139
GenBank mRNA AK022660.1 - 7 - 8 724 139
AK056761.1 - 9 - 10 1626 139
AK298750.1 - 6 - 7 626 139
AK303639.1 - 7 - 8 734 139
BC018720.1 - 9 - 10 938 139
CR457290.1 - 5 - 6 490 139

View Full Product Details