The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321135.1 | NP_001308064.1 | 12 - 13 | 1423 | 80 |
NM_001321136.1 | NP_001308065.1 | 14 - 15 | 1699 | 80 | |
NM_001321137.1 | NP_001308066.1 | 14 - 15 | 1296 | 80 | |
NM_003978.4 | NP_003969.2 | 13 - 14 | 1480 | 80 | |
NR_135552.1 | 10 - 11 | 1212 | 80 | ||
XM_006720737.2 | XP_006720800.1 | 13 - 14 | 1212 | 80 | |
XM_011522164.1 | XP_011520466.1 | 14 - 15 | 1166 | 80 | |
XM_011522165.2 | XP_011520467.1 | 13 - 14 | 1021 | 80 | |
XM_011522168.2 | XP_011520470.1 | 14 - 15 | 1498 | 80 | |
GenBank mRNA | AB451310.1 | - | 13 - 14 | 991 | 80 |
AB451440.1 | - | 13 - 14 | 991 | 80 | |
AF038602.1 | - | 12 - 13 | 934 | 80 | |
AF038603.1 | - | 13 - 14 | 991 | 80 | |
AK308462.1 | - | 14 - 15 | 1330 | 80 | |
AK309856.1 | - | 14 - 15 | 1662 | 80 | |
AL110110.1 | - | 2 - 3 | 140 | 80 | |
BC008602.1 | - | 13 - 14 | 1352 | 80 | |
KU178458.1 | - | 13 - 14 | 991 | 80 | |
KU178459.1 | - | 10 - 11 | 723 | 80 | |
U94778.1 | - | 13 - 14 | 1207 | 80 |