The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016219.4 | NP_057303.2 | 1 - 2 | 299 | 125 |
NR_045720.1 | 1 - 2 | 299 | 125 | ||
NR_045721.1 | 1 - 2 | 299 | 125 | ||
XM_006716945.3 | XP_006717008.1 | 1 - 2 | 364 | 125 | |
XM_017014239.1 | XP_016869728.1 | 1 - 2 | 145 | 125 | |
GenBank mRNA | AF145732.1 | - | 1 - 2 | 216 | 125 |
AF148509.1 | - | 1 - 2 | 276 | 125 | |
AK298480.1 | - | 1 - 2 | 145 | 125 | |
AK299043.1 | - | 1 - 2 | 146 | 125 | |
AK309808.1 | - | 1 - 2 | 145 | 125 | |
AK315797.1 | - | 1 - 2 | 241 | 125 | |
AY358465.1 | - | 1 - 2 | 248 | 125 | |
BC002953.1 | - | 1 - 2 | 232 | 125 | |
BC006079.1 | - | 1 - 2 | 241 | 125 | |
BM848429.1 | - | 1 - 2 | 298 | 125 | |
CR749534.1 | - | 1 - 2 | 141 | 125 | |
DA763005.1 | - | 1 - 2 | 261 | 125 |