The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016219.4 | NP_057303.2 | 8 - 9 | 1329 | 68 |
NR_045720.1 | 8 - 9 | 1329 | 68 | ||
NR_045721.1 | 9 - 10 | 1460 | 68 | ||
XM_006716945.3 | XP_006717008.1 | 8 - 9 | 1394 | 68 | |
XM_017014239.1 | XP_016869728.1 | 9 - 10 | 1306 | 68 | |
GenBank mRNA | AF145732.1 | - | 8 - 9 | 1246 | 68 |
AF148509.1 | - | 8 - 9 | 1306 | 68 | |
AK074699.1 | - | 6 - 7 | 904 | 68 | |
AK130143.1 | - | 3 - 4 | 473 | 68 | |
AK298480.1 | - | 9 - 10 | 1306 | 68 | |
AK299043.1 | - | 8 - 9 | 1176 | 68 | |
AK315797.1 | - | 8 - 9 | 1271 | 68 | |
AL110221.1 | - | 1 - 2 | 51 | 68 | |
AY358465.1 | - | 8 - 9 | 1278 | 68 | |
BC002953.1 | - | 8 - 9 | 1262 | 68 | |
BC006079.1 | - | 8 - 9 | 1271 | 68 |