The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001326381.1 | NP_001313310.1 | 1 - 2 | 405 | 61 |
NM_014320.2 | NP_055135.1 | 1 - 2 | 405 | 61 | |
XM_017010631.1 | XP_016866120.1 | 1 - 2 | 378 | 61 | |
GenBank mRNA | AF117616.1 | - | 1 - 2 | 383 | 61 |
AK057928.1 | - | 1 - 2 | 320 | 61 | |
AY427823.1 | - | 1 - 2 | 287 | 61 | |
BC008205.1 | - | 1 - 2 | 269 | 61 | |
BC010290.1 | - | 1 - 2 | 282 | 61 | |
BC016806.1 | - | 1 - 2 | 385 | 61 | |
BC016807.1 | - | 1 - 2 | 385 | 61 | |
BC093037.1 | - | 1 - 2 | 268 | 61 | |
BF974311.1 | - | - | 405 | 61 |