Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP2R5D
Entrez Gene ID
5528
Gene Name
protein phosphatase 2 regulatory subunit B'delta
Gene Aliases
B56D, MRD35
Chromosome Location
Chr.6: 42984499 - 43012345 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270476.1 NP_001257405.1 15 - 16 1868 66
NM_006245.3 NP_006236.1 15 - 16 1851 66
NM_180976.2 NP_851307.1 15 - 16 1755 66
NM_180977.2 NP_851308.1 13 - 14 1533 66
GenBank mRNA AB000634.1 - 15 - 16 1686 66
AB000635.1 - 13 - 14 1380 66
AB209861.1 - 12 - 13 1376 66
AB451342.1 - 15 - 16 1672 66
AB451357.1 - 15 - 16 1672 66
AK122701.1 - 15 - 16 1745 66
AK290604.1 - 15 - 16 1664 66
AK291797.1 - 15 - 16 1759 66
AK299696.1 - 15 - 16 1724 66
BC001095.2 - 15 - 16 1735 66
BC001175.1 - 15 - 16 1662 66
BC010692.1 - 15 - 16 1745 66
D78360.1 - 15 - 16 1653 66
L76702.1 - 15 - 16 1859 66

View Full Product Details