The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002296.3 | NP_002287.2 | 5 - 6 | 819 | 65 |
NM_194442.2 | NP_919424.1 | 5 - 6 | 809 | 65 | |
XM_005273125.2 | XP_005273182.1 | 5 - 6 | 775 | 65 | |
XM_011544185.2 | XP_011542487.1 | 5 - 6 | 1221 | 65 | |
GenBank mRNA | AB209514.1 | - | 5 - 6 | 726 | 65 |
AK222834.1 | - | 5 - 6 | 706 | 65 | |
AK303589.1 | - | 5 - 6 | 725 | 65 | |
AK312258.1 | - | 5 - 6 | 760 | 65 | |
BC020079.1 | - | 5 - 6 | 741 | 65 | |
L25931.1 | - | 5 - 6 | 720 | 65 |