The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135191.1 | NP_001128663.1 | 20 - 21 | 2358 | 91 |
NM_003887.2 | NP_003878.1 | 20 - 21 | 2358 | 91 | |
XM_006711898.1 | XP_006711961.1 | 21 - 22 | 2120 | 91 | |
XM_006711901.3 | XP_006711964.1 | 21 - 22 | 1970 | 91 | |
XM_006711902.1 | XP_006711965.1 | 21 - 22 | 2120 | 91 | |
XM_011510403.2 | XP_011508705.1 | 21 - 22 | 2141 | 91 | |
XM_011510404.2 | XP_011508706.1 | 20 - 21 | 2131 | 91 | |
XM_011510405.2 | XP_011508707.1 | 20 - 21 | 2067 | 91 | |
XM_011510406.2 | XP_011508708.1 | 22 - 23 | 2074 | 91 | |
XM_011510407.2 | XP_011508709.1 | 21 - 22 | 2141 | 91 | |
XM_011510408.2 | XP_011508710.1 | 21 - 22 | 2144 | 91 | |
XM_011510409.1 | XP_011508711.1 | 21 - 22 | 2012 | 91 | |
GenBank mRNA | AB007860.1 | - | 20 - 21 | 2358 | 91 |
BC063308.1 | - | 20 - 21 | 2311 | 91 |