The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135191.1 | NP_001128663.1 | 2 - 3 | 542 | 94 |
NM_003887.2 | NP_003878.1 | 2 - 3 | 542 | 94 | |
XM_006711898.1 | XP_006711961.1 | 2 - 3 | 295 | 94 | |
XM_006711901.3 | XP_006711964.1 | 2 - 3 | 145 | 94 | |
XM_006711902.1 | XP_006711965.1 | 2 - 3 | 295 | 94 | |
XM_011510403.2 | XP_011508705.1 | 2 - 3 | 316 | 94 | |
XM_011510404.2 | XP_011508706.1 | 2 - 3 | 315 | 94 | |
XM_011510405.2 | XP_011508707.1 | 2 - 3 | 317 | 94 | |
XM_011510406.2 | XP_011508708.1 | 3 - 4 | 249 | 94 | |
XM_011510407.2 | XP_011508709.1 | 2 - 3 | 316 | 94 | |
XM_011510408.2 | XP_011508710.1 | 2 - 3 | 319 | 94 | |
GenBank mRNA | AB007860.1 | - | 2 - 3 | 542 | 94 |
BC063308.1 | - | 2 - 3 | 495 | 94 |