Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C3P1
Entrez Gene ID
388503
Gene Name
complement component 3 precursor pseudogene
Gene Aliases
CPLP
Chromosome Location
Chr.19: 10041356 - 10074138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027300.2 2 - 3 235 65
GenBank mRNA DT932550.1 - 2 - 3 65

View Full Product Details