The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027300.2 | 8 - 9 | 1240 | 72 | |
GenBank mRNA | AK131489.1 | - | 6 - 7 | 1005 | 72 |