The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032595.4 | NP_115984.3 | 3 - 4 | 1630 | 117 |
XM_011525381.2 | XP_011523683.1 | 3 - 4 | 2140 | 117 | |
GenBank mRNA | AJ401189.1 | - | 3 - 4 | 1620 | 117 |
AK054907.1 | - | 1 - 2 | 103 | 117 | |
AL713642.1 | - | 3 - 4 | 437 | 117 | |
BC016162.1 | - | 1 - 2 | 117 | 117 |