The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199851.1 | NP_001186780.1 | 1 - 2 | 85 | 112 |
XM_017002292.1 | XP_016857781.1 | 2 - 3 | 170 | 112 | |
XM_017002295.1 | XP_016857784.1 | 2 - 3 | 170 | 112 | |
GenBank mRNA | AK298201.1 | - | 1 - 2 | 93 | 112 |