The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288629.1 | NP_001275558.1 | 2 - 3 | 405 | 99 |
NM_001288630.1 | NP_001275559.1 | 2 - 3 | 405 | 99 | |
NM_004440.3 | NP_004431.1 | 2 - 3 | 405 | 99 | |
XM_005248669.2 | XP_005248726.1 | 2 - 3 | 462 | 99 | |
XM_005248671.2 | XP_005248728.1 | 2 - 3 | 462 | 99 | |
XM_017010365.1 | XP_016865854.1 | 2 - 3 | 462 | 99 | |
XM_017010366.1 | XP_016865855.1 | 2 - 3 | 462 | 99 | |
GenBank mRNA | AB209269.1 | - | 2 - 3 | 392 | 99 |
AK313529.1 | - | 2 - 3 | 377 | 99 | |
BC027940.1 | - | 2 - 3 | 370 | 99 | |
BC126125.1 | - | 2 - 3 | 279 | 99 | |
BC126151.1 | - | 2 - 3 | 279 | 99 | |
BC143857.1 | - | 2 - 3 | 279 | 99 | |
BC143858.1 | - | 2 - 3 | 279 | 99 | |
DA503795.1 | - | 2 - 3 | 405 | 99 | |
L36642.1 | - | 2 - 3 | 349 | 99 |