Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDLBP
Entrez Gene ID
3069
Gene Name
high density lipoprotein binding protein
Gene Aliases
HBP, PRO2900, VGL
Chromosome Location
Chr.2: 241227264 - 241315842 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243900.2 NP_001230829.1 11 - 12 1530 69
NM_001320965.1 NP_001307894.1 11 - 12 1671 69
NM_001320966.1 NP_001307895.1 11 - 12 1529 69
NM_001320967.1 NP_001307896.1 11 - 12 1606 69
NM_005336.5 NP_005327.1 11 - 12 1753 69
NM_203346.4 NP_976221.1 11 - 12 1606 69
XM_005247002.3 XP_005247059.2 11 - 12 1560 69
XM_005247003.4 XP_005247060.2 11 - 12 1605 69
XM_006712475.3 XP_006712538.1 13 - 14 1809 69
XM_011511058.2 XP_011509360.1 13 - 14 1973 69
XM_011511060.2 XP_011509362.1 12 - 13 1821 69
XM_017003940.1 XP_016859429.1 12 - 13 1631 69
XM_017003941.1 XP_016859430.1 11 - 12 1748 69
XM_017003942.1 XP_016859431.1 11 - 12 1631 69
GenBank mRNA AK127833.1 - 5 - 6 710 69
AK300312.1 - 11 - 12 1475 69
AK312335.1 - 11 - 12 1583 69
BC001179.1 - 11 - 12 1579 69
BC014305.2 - 3 - 4 268 69
BX647759.1 - 11 - 12 1614 69
M64098.1 - 11 - 12 1532 69

View Full Product Details