The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243900.2 | NP_001230829.1 | 17 - 18 | 2324 | 76 |
NM_001320965.1 | NP_001307894.1 | 17 - 18 | 2465 | 76 | |
NM_001320966.1 | NP_001307895.1 | 17 - 18 | 2323 | 76 | |
NM_001320967.1 | NP_001307896.1 | 17 - 18 | 2400 | 76 | |
NM_005336.5 | NP_005327.1 | 17 - 18 | 2547 | 76 | |
NM_203346.4 | NP_976221.1 | 17 - 18 | 2400 | 76 | |
XM_005247002.3 | XP_005247059.2 | 17 - 18 | 2354 | 76 | |
XM_005247003.4 | XP_005247060.2 | 17 - 18 | 2399 | 76 | |
XM_006712475.3 | XP_006712538.1 | 19 - 20 | 2603 | 76 | |
XM_011511058.2 | XP_011509360.1 | 19 - 20 | 2767 | 76 | |
XM_011511060.2 | XP_011509362.1 | 18 - 19 | 2615 | 76 | |
XM_017003940.1 | XP_016859429.1 | 18 - 19 | 2425 | 76 | |
XM_017003941.1 | XP_016859430.1 | 17 - 18 | 2542 | 76 | |
XM_017003942.1 | XP_016859431.1 | 17 - 18 | 2425 | 76 | |
GenBank mRNA | AK127833.1 | - | 11 - 12 | 1504 | 76 |
AK131375.1 | - | 6 - 7 | 1757 | 76 | |
AK300312.1 | - | 17 - 18 | 2269 | 76 | |
AK312335.1 | - | 17 - 18 | 2377 | 76 | |
BC001179.1 | - | 17 - 18 | 2373 | 76 | |
BC014305.2 | - | 9 - 10 | 1062 | 76 | |
BC113120.1 | - | 1 - 2 | 137 | 76 | |
M64098.1 | - | 17 - 18 | 2326 | 76 |