The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243900.2 | NP_001230829.1 | 18 - 19 | 2541 | 80 |
NM_001320965.1 | NP_001307894.1 | 18 - 19 | 2682 | 80 | |
NM_001320966.1 | NP_001307895.1 | 18 - 19 | 2540 | 80 | |
NM_001320967.1 | NP_001307896.1 | 18 - 19 | 2617 | 80 | |
NM_005336.5 | NP_005327.1 | 18 - 19 | 2764 | 80 | |
NM_203346.4 | NP_976221.1 | 18 - 19 | 2617 | 80 | |
XM_005247002.3 | XP_005247059.2 | 18 - 19 | 2571 | 80 | |
XM_005247003.4 | XP_005247060.2 | 18 - 19 | 2616 | 80 | |
XM_006712475.3 | XP_006712538.1 | 20 - 21 | 2820 | 80 | |
XM_011511058.2 | XP_011509360.1 | 20 - 21 | 2984 | 80 | |
XM_011511060.2 | XP_011509362.1 | 19 - 20 | 2832 | 80 | |
XM_017003940.1 | XP_016859429.1 | 19 - 20 | 2642 | 80 | |
XM_017003941.1 | XP_016859430.1 | 18 - 19 | 2759 | 80 | |
XM_017003942.1 | XP_016859431.1 | 18 - 19 | 2642 | 80 | |
GenBank mRNA | AK127833.1 | - | 12 - 13 | 1721 | 80 |
AK131375.1 | - | 7 - 8 | 1974 | 80 | |
AK296684.1 | - | 12 - 13 | 1562 | 80 | |
AK300312.1 | - | 18 - 19 | 2486 | 80 | |
AK312335.1 | - | 18 - 19 | 2594 | 80 | |
BC001179.1 | - | 18 - 19 | 2590 | 80 | |
BC014305.2 | - | 10 - 11 | 1279 | 80 | |
BC113120.1 | - | 2 - 3 | 354 | 80 | |
BX647759.1 | - | 18 - 19 | 2625 | 80 | |
M64098.1 | - | 18 - 19 | 2543 | 80 |