The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018534.3 | NP_061004.3 | 14 - 15 | 3199 | 110 |
NM_201267.1 | NP_957719.1 | 14 - 15 | 3199 | 95 | |
XM_017005186.1 | XP_016860675.1 | 15 - 16 | 3064 | 95 | |
GenBank mRNA | AF280544.1 | - | 14 - 15 | 2408 | 95 |
AF280545.1 | - | 14 - 15 | 2408 | 110 | |
AK307367.1 | - | 2 - 3 | 166 | 95 | |
BC101525.1 | - | 14 - 15 | 2439 | 95 | |
BC104770.1 | - | 14 - 15 | 2439 | 95 | |
BC143237.1 | - | 14 - 15 | 2429 | 110 | |
BC143238.1 | - | 14 - 15 | 2439 | 95 |